EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Structures of deepoxytrichothecene metabolites from 3'-hydroxy HT-2 toxin and T-2 tetraol in rats.
PMID 4073895
Abstract
3'-Hydroxy HT-2 toxin and T-2 tetraol, in vivo metabolites of T-2 toxin, were orally administered to Wistar rats, and four metabolites having a trichothec-9,12-diene nucleus, which were termed deepoxytrichothecenes, were newly found in the excreta. Their structures were confirmed as 3'-hydroxy-deepoxy HT-2, 3'-hydroxy-deepoxy T-2 triol, 15-acetyl-deepoxy T-2 tetraol, and deepoxy T-2 tetraol on the basis of mass and nuclear magnetic resonance spectroscopy. Resolution of T-2 metabolites and corresponding deepoxytrichothecenes by gas-liquid and thin-layer chromatography was also described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32914
T-2-Tetraol solution, 50 μg/mL in acetonitrile, analytical standard
C15H22O6