EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Effect of perforated plastic row covers on residues of the herbicide DCPA in soil and broccoli.
PMID 4074938
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45397
Chlorthal-dimethyl, PESTANAL®, analytical standard
C10H6Cl4O4