EMAIL THIS PAGE TO A FRIEND
Drug intelligence & clinical pharmacy
Gitoxin--a challenger for digoxin?
PMID 4085356
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
64107
Gitoxin, ≥99% (HPLC)
C41H64O14