EMAIL THIS PAGE TO A FRIEND
Nucleic acids symposium series
Reaction of nucleic acids bases and their derivatives with peroxodisulfate ion.
PMID 4088884
Abstract
Reaction with peroxodisulfate ion was investigated, that is, reaction of 1,3-dimethyluracil, 1,3-dimethylthymine, and caffeine with carbon radicals formed from decarboxylation of carboxylic acids, oxidation of the methyl group at 5-position of thymines, and halogenation of nucleic acids bases and their derivatives with alkali halides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
349801
1,3-Dimethyluracil, 99%
C6H8N2O2