EMAIL THIS PAGE TO A FRIEND
Journal of veterinary pharmacology and therapeutics
Thiabendazole and 5-hydroxythiabendazole in the plasma of sheep.
PMID 4094032
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33818
5-Hydroxythiabendazole, PESTANAL®, analytical standard
C10H7N3OS