EMAIL THIS PAGE TO A FRIEND
Vestnik khirurgii imeni I. I. Grekova
[Effect of various cardiac glycosides on central hemodynamics after operations on the abdominal organs].
PMID 4095850
Abstract
The authors estimated the influence of corglycon, strophanthin and digoxin on the central hemodynamics indices. Corglycon is recommended for use in urgent situations, while digoxin should be used for prophylactics of cardiac insufficiency.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9140
Convallatoxin, ≥65%
C29H42O10