EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
[Derivatives of succinic acid. II. Synthesis and hypoglycemic properties of various derivatives of N-(benzenesulfonyl)succinamide].
PMID 4096250
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
124753
Succinamide, 98%
C4H8N2O2