EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Immunoglobulin and theta-bearing murine leukemias and lymphomas.
PMID 4112916
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
EL4 Cell Line murine, Lymphoblast from blood, 85023105