EMAIL THIS PAGE TO A FRIEND
Biochemistry
Purification and properties of beta-glucuronidase from human placenta.
PMID 413568
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73677
4-Nitrophenyl β-D-glucuronide, ≥99.0% (TLC)
C12H13NO9