EMAIL THIS PAGE TO A FRIEND
Cancer research
Reaction of drugs with nitrous acid as a source of carcinogenic nitrosamines.
PMID 4148978
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
109428
4-Hydroxyantipyrine, 99%
C11H12N2O2