EMAIL THIS PAGE TO A FRIEND
Enzyme
Semiautomated enzymic microassay for plasma L-alanine.
PMID 41707
Abstract
Plasma L-alanine is deaminated by bacterial alanine-dehydrogenase; the resulting ammonia is dialyzed out and measured by use of a continuous flow phenol-hypochlorite colorimetric microassay. Concentrations in the range of 10-1,000 mumol alanine/l can be determined in a 50-microliter sample. The optimal conditions for the assay are specified. Study of the analytical qualities of the technique shows high specificity, good reproducibility , and a detection limit of 6 mumol/l. Usual values in human plasma from arterial or venous blood are respectively 296 +/- 166 and 376 +/- 214 mumol alanine/l (x +/- 2 SD). The usual values in rats are close to those found in man.