EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
beta-Galactosidase from rat mammary gland. Its purification, properties, and role in the biosynthesis of 6beta-O-D-galactopyranosyl myo-inositol.
PMID 418065