EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Mixed culture cytopathogenicity: a new test for growth of murine leukemia viruses in tissue culture.
PMID 4186808
Abstract
Tissue culture cells of a Rous virus-induced rat tumor undergo syncytium formation when placed in contact with mouse embryo cell cultures infected with murine leukemia viruses. This phenomenon can be used as a cytopathic end point for isolating and titrating these viruses in tissue culture. The principle should be applicable to detection of leukemia viruses of other species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
XC
details
Y1