EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Stabilization of Bacillus subtilis -amylase by amino group acylation.
PMID 4197008
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
20430
Boc-OSu, ≥98.0% (CHN)
C9H13NO5