EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A rapid electrophoretic procedure for the detection of SDS-released oncorna-viral RNA using polyacrylamide-agarose gels.
PMID 4201521
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L3771
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
C12H25NaO4S
L4509
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
C12H25NaO4S
L6026
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
C12H25NaO4S
436143
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
C12H25NaO4S
71725
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
C12H25NaO4S
71726
Sodium dodecyl sulfate, for ion pair chromatography, LiChropur, ≥99.0%
C12H25NaO4S
L5750
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
C12H25NaO4S
75746
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
C12H25NaO4S
71729
Sodium dodecyl sulfate, ≥98.0% (GC)
C12H25NaO4S
62862
Sodium dodecyl sulfate, ≥90%
C12H25NaO4S
74255
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
C12H25NaO4S
71717
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
C12H25NaO4S
RDD021
Sodium dodecyl sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
C12H25NaO4S
Y0000620
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
C12H25NaO4S
1614363
Sodium lauryl sulfate, United States Pharmacopeia (USP) Reference Standard
C12H25NaO4S
PHR1949
Sodium Lauryl Sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
C12H25NaO4S