EMAIL THIS PAGE TO A FRIEND
The Medical clinics of North America
Griseofulvin.
PMID 4248513
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0600000
Griseofulvin, European Pharmacopoeia (EP) Reference Standard
C17H17ClO6