EMAIL THIS PAGE TO A FRIEND
Toxicology and applied pharmacology
Thiodiglycolic acid: a major metabolite of bis(2-chloroethyl)ether.
PMID 425117
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35660
Bis(2-chloroethyl) ether, puriss., ≥99.0% (GC)
C4H8Cl2O