EMAIL THIS PAGE TO A FRIEND
South African medical journal = Suid-Afrikaanse tydskrif vir geneeskunde
Significance of the porphyrin precursors delta-aminolaevulinic acid (ALA) and porphobilinogen (PBG) in the acute attack of porphyria.
PMID 4270402
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1134
Porphobilinogen, powder
C10H14N2O4