EMAIL THIS PAGE TO A FRIEND
Virology
Antigenic and cultural properties of cells doubly transformed by polyoma virus and SV40.
PMID 4284655
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
3T3 Swiss Albino, 85022108
details
SV3T3B, 85011409