EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
The subcellular distribution of enzymes in type II glycogenosis and the occurrence of an oligo-alpha-1,4-glucan glucohydrolase in human tissues.
PMID 4286143
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
10113 Amyloglucosidase from Aspergillus niger, powder, white, ~120 U/mg