EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Microenzymic method for the quantitative analysis of acetyl groups in proteins and peptides.
PMID 4292707
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
268925
Butyl ethyl ether, 99%
C6H14O