EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Mechanism of the salicylate hydroxylase reaction. 3. Characterization and reactivity of chemically or photochemically reduced enzyme-flavin.
PMID 4309912