EMAIL THIS PAGE TO A FRIEND
International journal of cancer
Isolation of murine sarcoma virus-transformed mouse cells which are negative for leukemia virus from agar suspension cultures.
PMID 4319233
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
3T3-MSV, 90092701