EMAIL THIS PAGE TO A FRIEND
The Journal of clinical endocrinology and metabolism
Use of benzamidine as a proteolytic inhibitor in the radioimmunoassay of glucagon in plasma.
PMID 4340616
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
12072
Benzamidine, ≥95.0%
C7H8N2