EMAIL THIS PAGE TO A FRIEND
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Periodate-lysine-paraformaldehyde fixative. A new fixation for immunoelectron microscopy.
PMID 4374474
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
158127
Paraformaldehyde, powder, 95%
CH2O ]n