EMAIL THIS PAGE TO A FRIEND
Endocrinology
Studies on interstitial cells in tissue culture: steroid biosynthesis in monolayers of mouse testicular interstitial cells.
PMID 4382264
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
I-10, 89031605