EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Oxidation of 1,5-diphenylcarbazide as a measure of photosystem 2 activity in subchloroplast fragments.
PMID 4390522
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
259225
1,5-Diphenylcarbazide, ACS reagent
C13H14N4O