EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Synthesis of 1-p-chlorobenzyl-7-azaindole-3- -piperidylmethanol as a potential antimalarial agent.
PMID 4400149
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
694681
7-Azaindole-3-carboxaldehyde, 97%
C8H6N2O