EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacy and pharmacology
Biological properties of carrageenan.
PMID 4402944
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1867 Carrageenan, Non-gelling, Mixture of κ and λ carrageenans