EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
2-Amino-2-methyl-1-propanol as the alkalizing agent in an improved continuous-flow cresolphthalein complexone procedure for calcium in serum.
PMID 4421368