EMAIL THIS PAGE TO A FRIEND
Polish journal of pharmacology and pharmacy
Nicotinic acid derivatives. VI. Some transformations of 2-chloro-6-methylnicotinic acid.
PMID 4427816
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357847
2-Chloro-6-methylpyridine-3-carboxylic acid, 97%
C7H6ClNO2