EMAIL THIS PAGE TO A FRIEND
Biochemistry
Calcium uptake and membrane potential in mitochondria.
PMID 4429666
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1779 Nigericin sodium salt Ready Made Solution, 5 mg/mL (DMSO:ethanol 1:1)
C40H67NaO11