EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Uridine diphosphate glucose pyrophosphorylase. Crystallization and properties of the enzyme from rabbit liver and species comparisons.
PMID 4436332