EMAIL THIS PAGE TO A FRIEND
La Prensa medica mexicana
[Central venous pressure in the acutely ill patient. Technic, interpretation and complications].
PMID 4457876
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
PA-1