EMAIL THIS PAGE TO A FRIEND
Endocrinology
Zearalenones: characterization of the estrogenic potencies and receptor interactions of a series of fungal beta-resorcylic acid lactones.
PMID 446414
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Z0417
β-Zearalanol, ~98% (HPLC)
C18H26O5