EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Affinity chromatography of carboxypeptidase B.
PMID 4476043
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
15395
Boc-6-Ahx-OH, ≥99.0% (T)
C11H21NO4