EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
The site of diazoacetyl inhibitor attachment to acid proteinase of Aspergillus awamori--an analog of penicillopepsin and pepsin.
PMID 4565799
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2143 Protease from Aspergillus saitoi, Type XIII, ≥0.6 unit/mg solid