EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
The stereochemical configuration of the lactoyl group of S-lactoylglutathionine formed by the action of glyoxalase I from porcine erythrocytes and yeast.
PMID 4574550