EMAIL THIS PAGE TO A FRIEND
Quarterly reviews of biophysics
The lesions produced by ultraviolet light in DNA containing 5-bromouracil.
PMID 4579676
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
852473
5-Bromouracil, 98%
C4H3BrN2O2