EMAIL THIS PAGE TO A FRIEND
Chemico-biological interactions
Alkylation of transfer RNA by N-methyl-N-nitrosourea and N-ethyl-N-nitrosourea.
PMID 4583979
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
363057
6-O-Methylguanine, 97%
C6H7N5O