EMAIL THIS PAGE TO A FRIEND
Fortschritte der Chemie organischer Naturstoffe = Progress in the chemistry of organic natural products. Progres dans la chimie des substances organiques naturelles
[Chemistry and biology of saponins].
PMID 4617698