EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Ornithine transcarbamylase from Streptococcus faecalis and bovine liver. 3. Effects of chemical modifications of specific residues on ligand binding and enzymatic activity.
PMID 4622305