EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of immunology
Separation of cells according to surface antigens by the use of antibody-coated columns. Fractionation of cells carrying immunoglobulins and blood group antigen.
PMID 4627539