EMAIL THIS PAGE TO A FRIEND
The Analyst
The quantitative extraction and gas-liquid chromatographic determination of organic acids in urine.
PMID 4651954
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33035-U BSA Derivatization Grade, derivatization grade, Silylation reagent