EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Urinary guanidinoacetate-guanidinosuccinate ratio: an indicator of kidney dysfunction.
PMID 4689101
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
251976
2,4-Dichloro-1-naphthol, 95%
C10H6Cl2O