EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
A sensitive gas chromatographic procedure for the estimation of noradrenaline, dopamine and 5-hydroxytryptamine in rat brain.
PMID 4691879
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91719
Trifluoroacetic anhydride, for GC derivatization, ≥99.0% (GC)
C4F6O3