EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
In vivo studies on the introduction of the 4-t-double bond of the sphingenine moiety of rat brain ceramides.
PMID 4708093
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3314
D-erythro-Dihydrosphingosine, ≥98%
C18H39NO2