EMAIL THIS PAGE TO A FRIEND
Cancer research
N-demethylation of the antineoplastic agent hexamethylmelamine by rats and man.
PMID 4748436
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
126365
Dimethylamine hydrochloride, 99%
C2H7N · HCl