EMAIL THIS PAGE TO A FRIEND
Biochemistry
High-yield cleavage of tryptophanyl peptide bonds by o-iodosobenzoic acid.
PMID 476090
Abstract
A new procedure to cleave tryptophanyl peptide bonds in high yield is reported. The method involves treatment of the S-alkylated protein with o-iodosobenzoic acid. The procedure is highly selective for tryptophan and does not modify tyrosine or histidine, but may convert methionine to its sulfoxide derivative. The yields in the cleavage are 70--100%. Tryptophanyl bonds to alanine, glycine, serine, threonine, glutamine, arginine, and S-(pyridylethyl)cysteine are split in nearly quantitative yield, while those preceding isoleucine or valine are split in approximately 70% yield in the proteins examined in this work. The chemical mechanism for tryptophanyl bond cleavage has not been defined, but it is likely that oxidation of the indole ring occurs during the reaction with o-iodosobenzoic acid. Some problems with the quality of commercial preparations of the reagent are discussed.