EMAIL THIS PAGE TO A FRIEND
Acta pharmaceutica Suecica
Microsomal N-hydroxylation of phenylalkylamines. Identification of N-hydroxylated phenylalkylamines as their trimethylsilyl derivatives by GC-Ms.
PMID 4773116